2-Chloro-3-ethyl-quinoline

Lieferant: Apollo Scientific

OR309257-5G
APOSOR309257-5GEA 172 EUR
APOSOR309257-5G
2-Chloro-3-ethyl-quinoline
2-Chloro-3-ethyl-quinoline
Formel: C₁₁H₁₀ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD02684198
CAS-Nummer: 67525-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR