2-Chloro-6-ethoxy-3-phenylquinoline

Lieferant: Apollo Scientific

OR309259-1G
APOSOR309259-1GEA 359 EUR
APOSOR309259-1G
2-Chloro-6-ethoxy-3-phenylquinoline
2-Chloro-6-ethoxy-3-phenylquinoline
Formel: C₁₇H₁₄ClNO
Molecular Weight: 283,752 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998012
CAS-Nummer: 1031928-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR