2-Chloro-6-methoxy-3-phenylquinoline

Lieferant: Apollo Scientific

OR309261-1G
APOSOR309261-1GEA 325 EUR
APOSOR309261-1G
2-Chloro-6-methoxy-3-phenylquinoline
2-Chloro-6-methoxy-3-phenylquinoline
Formel: C₁₆H₁₂ClNO
Molecular Weight: 269,726 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998016
CAS-Nummer: 85274-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR