2-Chloroquinoline-3-methanol

Lieferant: Apollo Scientific

Synonyme: (2-Chloroquinolin-3-yl)methanol

OR309269-1G
APOSOR309269-1GEA 235 EUR
APOSOR309269-1G
2-Chloroquinoline-3-methanol
2-Chloroquinoline-3-methanol
Formel: C₁₀H₈ClNO
Molecular Weight: 193,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03694292
CAS-Nummer: 125917-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR