6-Bromo-2-chloro-3-ethylquinoline

Lieferant: Apollo Scientific

OR309270-1G
APOSOR309270-1GEA 272 EUR
APOSOR309270-1G
6-Bromo-2-chloro-3-ethylquinoline
6-Bromo-2-chloro-3-ethylquinoline
Formel: C₁₁H₉BrClN
Molecular Weight: 270,553 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09029946
CAS-Nummer: 409346-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR