6-Bromo-2-chloroquinoline-3-methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR309272-1G
APOSOR309272-1GEA
317
EUR
APOSOR309272-1G
6-Bromo-2-chloroquinoline-3-methanol
6-Bromo-2-chloroquinoline-3-methanol
Formel:
C₁₀H₇BrClNO Schmelzpunkt: 182…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09997953 CAS-Nummer: 1017464-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...