7-Bromo-2-chloro-3-ethylquinoline

Lieferant: Apollo Scientific

OR309273-1G
APOSOR309273-1GEA 401 EUR
APOSOR309273-1G
7-Bromo-2-chloro-3-ethylquinoline
7-Bromo-2-chloro-3-ethylquinoline
Formel: C₁₁H₉BrClN
Molecular Weight: 270,553 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997963
CAS-Nummer: 132118-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR