7-Bromo-2-chloro-3-phenylquinoline

Lieferant: Apollo Scientific

OR309274-1G
APOSOR309274-1GEA 542 EUR
APOSOR309274-1G
7-Bromo-2-chloro-3-phenylquinoline
7-Bromo-2-chloro-3-phenylquinoline
Formel: C₁₅H₉BrClN
Molecular Weight: 318,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997964
CAS-Nummer: 85274-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR