2-chloro-8-methylquinoline-3-carbaldehyde

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-8-methylquinoline-3-carboxaldehyde, 2-Chloro-3-formyl-8-methylquinoline, 2-Chloro-8-methylquinoline-3-carbaldehyde

OR309277-5G
APOSOR309277-5GEA 42.2 EUR
APOSOR309277-5G
2-chloro-8-methylquinoline-3-carbaldehyde
2-chloro-8-methylquinoline-3-carbaldehyde
Formel: C₁₁H₈ClNO
Molecular Weight: 205,641 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02227041
CAS-Nummer: 73568-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR