7-Bromo-4-hydroxy-2-methylquinoline
Lieferant: Apollo Scientific
OR309279-1G
APOSOR309279-1GEA
343
EUR
APOSOR309279-1G
7-Bromo-4-hydroxy-2-methylquinoline
7-Bromo-4-hydroxy-2-methylquinoline
Formel:
C₁₀H₈BrNO Molecular Weight: 238,081 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09997975 CAS-Nummer: 56716-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...