6-Bromo-4-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR309283-1G
APOSOR309283-1GEA 359 EUR
APOSOR309283-1G
6-Bromo-4-methylquinoline-3-carboxylic acid
6-Bromo-4-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈BrNO₂
Molecular Weight: 266,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052869
CAS-Nummer: 1095010-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR