7-Bromo-3-hydroxyquinoline-4-carboxylic acid
Lieferant: Apollo Scientific
OR309284-1G
APOSOR309284-1GEA
665
EUR
APOSOR309284-1G
7-Bromo-3-hydroxyquinoline-4-carboxylic acid
7-Bromo-3-hydroxyquinoline-4-carboxylic acid
Formel:
C₁₀H₆BrNO₃ Molecular Weight: 268,064 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10703487 CAS-Nummer: 1031929-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...