4-Bromo-7-methylquinoline
Lieferant: Apollo Scientific
OR309285-1G
APOSOR309285-1GEA
525
EUR
APOSOR309285-1G
4-Bromo-7-methylquinoline
4-Bromo-7-methylquinoline
Formel:
C₁₀H₈BrN Molecular Weight: 222,081 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08063191 CAS-Nummer: 1070879-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...