4-Bromo-5,7-dimethylquinoline

Lieferant: Apollo Scientific

OR309286-1G
APOSOR309286-1GEA 375 EUR
APOSOR309286-1G
4-Bromo-5,7-dimethylquinoline
4-Bromo-5,7-dimethylquinoline
Formel: C₁₁H₁₀BrN
Molecular Weight: 236,108 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505114
CAS-Nummer: 1070879-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR