4-Bromo-5,8-dichloroquinoline

Lieferant: Apollo Scientific

OR309287-1G
APOSOR309287-1GEA 498 EUR
APOSOR309287-1G
4-Bromo-5,8-dichloroquinoline
4-Bromo-5,8-dichloroquinoline
Formel: C₉H₄BrCl₂N
Molecular Weight: 276,945 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505120
CAS-Nummer: 1070879-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR