4-Bromo-6-ethyl-2-methylquinoline

Lieferant: Apollo Scientific

OR309288-1G
APOSOR309288-1GEA 375 EUR
APOSOR309288-1G
4-Bromo-6-ethyl-2-methylquinoline
4-Bromo-6-ethyl-2-methylquinoline
Formel: C₁₂H₁₂BrN
Molecular Weight: 250,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505128
CAS-Nummer: 1070879-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR