4-Bromo-6-chloro-2-methylquinoline
Lieferant: Apollo Scientific
OR309291-1G
APOSOR309291-1GEA
368
EUR
APOSOR309291-1G
4-Bromo-6-chloro-2-methylquinoline
4-Bromo-6-chloro-2-methylquinoline
Formel:
C₁₀H₇BrClN Molecular Weight: 256,526 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11505135 CAS-Nummer: 1070879-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...