4,6-Dibromo-2-methylquinoline
Lieferant: Apollo Scientific
OR309292-1G
APOSOR309292-1GEA
375
EUR
APOSOR309292-1G
4,6-Dibromo-2-methylquinoline
4,6-Dibromo-2-methylquinoline
Formel:
C₁₀H₇Br₂N Molecular Weight: 300,977 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261090 CAS-Nummer: 1070879-53-6 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...