4,8-Dibromo-2-methylquinoline

Lieferant: Apollo Scientific

OR309293-1G
APOSOR309293-1GEA 375 EUR
APOSOR309293-1G
4,8-Dibromo-2-methylquinoline
4,8-Dibromo-2-methylquinoline
Formel: C₁₀H₇Br₂N
Molecular Weight: 300,977 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505139
CAS-Nummer: 1070879-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR