4-Bromo-2,6,8-trimethylquinoline

Lieferant: Apollo Scientific

OR309294-1G
APOSOR309294-1GEA 409 EUR
APOSOR309294-1G
4-Bromo-2,6,8-trimethylquinoline
4-Bromo-2,6,8-trimethylquinoline
Formel: C₁₂H₁₂BrN
Molecular Weight: 250,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505145
CAS-Nummer: 1070879-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR