2-Hydrazino-6-methoxy-3-methylquinoline hydrochloride

Lieferant: Apollo Scientific

OR309296-1G
APOSOR309296-1GEA 368 EUR
APOSOR309296-1G
2-Hydrazino-6-methoxy-3-methylquinoline hydrochloride
2-Hydrazino-6-methoxy-3-methylquinoline hydrochloride
Formel: C₁₁H₁₃N₃O·ClH
Molecular Weight: 239.701 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505163
CAS-Nummer: 1172797-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR