4-Hydroxy-7-methoxy-2-phenylquinoline

Lieferant: Apollo Scientific

Synonyme: 7-Methoxy-2-phenylquinolin-4(1H)-one

OR309300-1G
APOSOR309300-1GEA 410 EUR
APOSOR309300-1G
4-Hydroxy-7-methoxy-2-phenylquinoline
4-Hydroxy-7-methoxy-2-phenylquinoline
Formel: C₁₆H₁₃NO₂
Molecular Weight: 251,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08274987
CAS-Nummer: 20430-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR