4-Hydroxy-8-methyl-2-propylquinoline

Lieferant: Apollo Scientific

Synonyme: 8-Methyl-2-propylquinolin-4-ol

OR309304-1G
APOSOR309304-1GEA 375 EUR
APOSOR309304-1G
4-Hydroxy-8-methyl-2-propylquinoline
4-Hydroxy-8-methyl-2-propylquinoline
Formel: C₁₃H₁₅NO
Molecular Weight: 201,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505250
CAS-Nummer: 1070879-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR