4-Hydroxy-6-methoxy-2-propylquinoline

Lieferant: Apollo Scientific

Synonyme: 6-Methoxy-2-propylquinolin-4-ol

OR309305-1G
APOSOR309305-1GEA 980 EUR
APOSOR309305-1G
4-Hydroxy-6-methoxy-2-propylquinoline
4-Hydroxy-6-methoxy-2-propylquinoline
Formel: C₁₃H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02684129
CAS-Nummer: 927800-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR