4-Hydroxy-7-methoxy-2-propylquinoline

Lieferant: Apollo Scientific

OR309306-1G
APOSOR309306-1GEA 383 EUR
APOSOR309306-1G
4-Hydroxy-7-methoxy-2-propylquinoline
4-Hydroxy-7-methoxy-2-propylquinoline
Formel: C₁₃H₁₅NO₂
Molecular Weight: 217,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505253
CAS-Nummer: 1070879-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR