4-Chloro-6,8-dimethyl-2-phenylquinoline

Lieferant: Apollo Scientific

OR309307-1G
APOSOR309307-1GEA 351 EUR
APOSOR309307-1G
4-Chloro-6,8-dimethyl-2-phenylquinoline
4-Chloro-6,8-dimethyl-2-phenylquinoline
Formel: C₁₇H₁₄ClN
Molecular Weight: 267,753 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674825
CAS-Nummer: 101602-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR