4-Bromo-6-methoxy-2-propylquinoline hydrobromide

Lieferant: Apollo Scientific

OR309308-1G
APOSOR309308-1GEA 525 EUR
APOSOR309308-1G
4-Bromo-6-methoxy-2-propylquinoline hydrobromide
4-Bromo-6-methoxy-2-propylquinoline hydrobromide
Formel: C₁₃H₁₄BrNO·BrH
Molecular Weight: 361.072 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674909
CAS-Nummer: 1204811-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR