5-Brom-2-nitroanisol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-methoxy-1-nitrobenzene

OR30931-10G OR30931-1G OR30931-5G
APOSOR30931-10GEA 29 EUR
APOSOR30931-10G APOSOR30931-1G APOSOR30931-5G
5-Brom-2-nitroanisol
5-Brom-2-nitroanisol
Formel: C₇H₆BrNO₃
Molecular Weight: 232.03 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09966092
CAS-Nummer: 103966-66-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR