7-Bromo-4-hydroxy-8-methylquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
OR309310-1G
APOSOR309310-1GEA
359
EUR
APOSOR309310-1G
7-Bromo-4-hydroxy-8-methylquinoline-3-carboxylic acid
7-Bromo-4-hydroxy-8-methylquinoline-3-carboxylic acid
Formel:
C₁₁H₈BrNO₃ Molecular Weight: 282,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12674984 CAS-Nummer: 1189107-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...