7-Bromo-4-hydroxy-8-methylquinoline
Lieferant: Apollo Scientific
OR309311-1G
APOSOR309311-1GEA
359
EUR
APOSOR309311-1G
7-Bromo-4-hydroxy-8-methylquinoline
7-Bromo-4-hydroxy-8-methylquinoline
Formel:
C₁₀H₈BrNO Molecular Weight: 238,081 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12674985 CAS-Nummer: 1189106-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...