8-Bromo-4-chloro-6-methylquinoline
Lieferant: Apollo Scientific
OR309312-1G
APOSOR309312-1GEA
433
EUR
APOSOR309312-1G
8-Bromo-4-chloro-6-methylquinoline
8-Bromo-4-chloro-6-methylquinoline
Formel:
C₁₀H₇BrClN Molecular Weight: 256,526 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12192763 CAS-Nummer: 1156602-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...