4-Bromo-6-chloro-8-methoxyquinoline
Lieferant: Apollo Scientific
OR309313-1G
APOSOR309313-1GEA
540
EUR
APOSOR309313-1G
4-Bromo-6-chloro-8-methoxyquinoline
4-Bromo-6-chloro-8-methoxyquinoline
Formel:
C₁₀H₇BrClNO Molecular Weight: 272,526 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12675014 CAS-Nummer: 1189107-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...