4-Bromo-8-chloro-5-methoxyquinoline
Lieferant: Apollo Scientific
OR309314-1G
APOSOR309314-1GEA
1360
EUR
APOSOR309314-1G
4-Bromo-8-chloro-5-methoxyquinoline
4-Bromo-8-chloro-5-methoxyquinoline
Formel:
C₁₀H₇BrClNO Molecular Weight: 272,526 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12675020 CAS-Nummer: 1189107-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...