8-Bromo-4-chloro-2,6-dimethylquinoline

Lieferant: Apollo Scientific

OR309316-1G
APOSOR309316-1GEA 0 EUR
APOSOR309316-1G
8-Bromo-4-chloro-2,6-dimethylquinoline
8-Bromo-4-chloro-2,6-dimethylquinoline
Formel: C₁₁H₉BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD12115058
CAS-Nummer: 1156275-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR