7-Bromo-8-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309317-1G
APOSOR309317-1GEA 375 EUR
APOSOR309317-1G
7-Bromo-8-methylquinoline-3-carboxylic acid
7-Bromo-8-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈BrNO₂
Molecular Weight: 266,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12675067
CAS-Nummer: 1189107-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR