7-Bromo-2,8-dimethylquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR309319-1G
APOSOR309319-1GEA
343
EUR
APOSOR309319-1G
7-Bromo-2,8-dimethylquinoline-3-carboxylic acid
7-Bromo-2,8-dimethylquinoline-3-carboxylic acid
Formel:
C₁₂H₁₀BrNO₂ Molecular Weight: 280,117 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12675072 CAS-Nummer: 1189106-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...