7-Bromo-2,8-dimethylquinoline

Lieferant: Apollo Scientific

OR309320-1G
APOSOR309320-1GEA 620 EUR
APOSOR309320-1G
7-Bromo-2,8-dimethylquinoline
7-Bromo-2,8-dimethylquinoline
Formel: C₁₁H₁₀BrN
Molecular Weight: 236.108 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12675073
CAS-Nummer: 1189106-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR