3-Bromo-4-hydroxyquinoline

Lieferant: Apollo Scientific

Synonyme: 3-Bromoquinolin-4-ol

OR309321-1G
APOSOR309321-1GEA 108 EUR
APOSOR309321-1G
3-Bromo-4-hydroxyquinoline
3-Bromo-4-hydroxyquinoline
Formel: C₉H₆BrNO
Molecular Weight: 224.054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234483
CAS-Nummer: 64965-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR