3,7-Dibromo-4-hydroxyquinoline

Lieferant: Apollo Scientific

OR309323-1G
APOSOR309323-1GEA 409 EUR
APOSOR309323-1G
3,7-Dibromo-4-hydroxyquinoline
3,7-Dibromo-4-hydroxyquinoline
Formel: C₉H₅Br₂NO
Molecular Weight: 302,95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192872
CAS-Nummer: 1203579-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR