3-Chloro-7-bromo-4-hydroxyquinoline

Lieferant: Apollo Scientific
Achtung

OR309324-1G
APOSOR309324-1GEA 426 EUR
APOSOR309324-1G
3-Chloro-7-bromo-4-hydroxyquinoline
3-Chloro-7-bromo-4-hydroxyquinoline
Formel: C₉H₅BrClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD13192873
CAS-Nummer: 1203579-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR