3,8-Dibromo-4-hydroxyquinoline

Lieferant: Apollo Scientific

OR309325-1G
APOSOR309325-1GEA 375 EUR
APOSOR309325-1G
3,8-Dibromo-4-hydroxyquinoline
3,8-Dibromo-4-hydroxyquinoline
Formel: C₉H₅Br₂NO
Molecular Weight: 302,95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192874
CAS-Nummer: 1204812-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR