3-Bromo-6,7-dimethyl-4-hydroxyquinoline

Lieferant: Apollo Scientific

OR309326-1G
APOSOR309326-1GEA 375 EUR
APOSOR309326-1G
3-Bromo-6,7-dimethyl-4-hydroxyquinoline
3-Bromo-6,7-dimethyl-4-hydroxyquinoline
Formel: C₁₁H₁₀BrNO
Molecular Weight: 252,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192891
CAS-Nummer: 1204811-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR