4-Bromo-6-methoxy-2-phenylquinoline
Lieferant: Apollo Scientific
OR309327-1G
APOSOR309327-1GEA
426
EUR
APOSOR309327-1G
4-Bromo-6-methoxy-2-phenylquinoline
4-Bromo-6-methoxy-2-phenylquinoline
Formel:
C₁₆H₁₂BrNO Molecular Weight: 314,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13192938 CAS-Nummer: 876491-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...