3-Bromo-4-chloro-6-methylquinoline

Lieferant: Apollo Scientific

OR309329-1G
APOSOR309329-1GEA 409 EUR
APOSOR309329-1G
3-Bromo-4-chloro-6-methylquinoline
3-Bromo-4-chloro-6-methylquinoline
Formel: C₁₀H₇BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD13192941
CAS-Nummer: 1204810-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR