3-Bromo-4-chloro-8-methylquinoline
Lieferant: Apollo Scientific
OR309330-1G
APOSOR309330-1GEA
0
EUR
APOSOR309330-1G
3-Bromo-4-chloro-8-methylquinoline
3-Bromo-4-chloro-8-methylquinoline
Formel:
C₁₀H₇BrClN Lagertemperatur: Raumtemperatur |
MDL:
MFCD13192945 CAS-Nummer: 1204810-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...