3-Bromo-4-chloro-6-methoxyquinoline

Lieferant: Apollo Scientific

OR309331-1G
APOSOR309331-1GEA 208 EUR
APOSOR309331-1G
3-Bromo-4-chloro-6-methoxyquinoline
3-Bromo-4-chloro-6-methoxyquinoline
Formel: C₁₀H₇BrClNO
Molecular Weight: 272,526 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192951
CAS-Nummer: 1203579-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR