3-Bromo-4-chloro-7-methoxyquinoline

Lieferant: Apollo Scientific
Achtung

OR309332-1G
APOSOR309332-1GEA 393 EUR
APOSOR309332-1G
3-Bromo-4-chloro-7-methoxyquinoline
3-Bromo-4-chloro-7-methoxyquinoline
Formel: C₁₀H₇BrClNO
Molecular Weight: 272,526 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192952
CAS-Nummer: 1203579-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR