3,4-Dichloro-7-methoxyquinoline

Lieferant: Apollo Scientific
Achtung

OR309333-1G
APOSOR309333-1GEA 409 EUR
APOSOR309333-1G
3,4-Dichloro-7-methoxyquinoline
3,4-Dichloro-7-methoxyquinoline
Formel: C₁₀H₇Cl₂NO
Molecular Weight: 228,075 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192953
CAS-Nummer: 1204810-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR