3-Bromo-4-chloro-8-methoxyquinoline

Lieferant: Apollo Scientific
Achtung

OR309334-1G
APOSOR309334-1GEA 375 EUR
APOSOR309334-1G
3-Bromo-4-chloro-8-methoxyquinoline
3-Bromo-4-chloro-8-methoxyquinoline
Formel: C₁₀H₇BrClNO
Molecular Weight: 272.526 g/mol
Schmelzpunkt: 129…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13192954
CAS-Nummer: 142781-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR