4-Chloro-3,6-dibromoquinoline

Lieferant: Apollo Scientific

OR309335-1G
APOSOR309335-1GEA 401 EUR
APOSOR309335-1G
4-Chloro-3,6-dibromoquinoline
4-Chloro-3,6-dibromoquinoline
Formel: C₉H₄Br₂ClN
Molecular Weight: 321,396 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437002
CAS-Nummer: 927801-19-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR